Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Flavin Containing Monooxygenase 3
- Antigen symbol:FMO3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2328
- Antigen synonyms:dJ127D3.1|FMOII|TMAU
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The Flavin containing monooxygenase family consists of five gene products, FMO1-5, that are major enzymatic oxidants involved in the metabolism of various therapeutics. Located in the liver, FMO3 is a hepatic microsomal enzyme that oxygenates soft nucleophiles such as secondary and tertiary amines. Through its N-oxygenase capabilities, FMO3 acts on a variety of xenobiotics to catalyze oxidative digestion. Defects in the FMO3 gene are the primary cause of trimethylaminuria (TMAuria), an inborn error of metabolism associated with a fishy body odor emitting from sweat, urine and breath. Genetic mutations in FMO3 lead to the N-oxidation of amino-trimethylamine derived from food products, thus producing the malodor associated with TMAuria.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FMO3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat