Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:F-box Protein 45
- Antigen symbol:FBXO45
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:200933
- Antigen synonyms:hFbxo45.|FBSP1_HUMAN|F box protein 45|F-box only protein 45|FBX45|fbxo45|F-box/SPRY domain-containing protein 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Belonging to the F-box family of proteins, FBXO45 is a 286 amino acid protein that contains one C-terminal F-box domain. F-box proteins are critical components of the SCF (Skp1-CUL-1-F-box protein)-type E3 ubiquitin ligase complex and are involved in substrate recognition and protein recruitment for ubiquitination. They are members of a larger family of proteins that are involved in the regulation of a wide variety of cellular mechanisms, including the cell cycle, the immune response, signaling cascades and developmental processes. They function by targeting proteins, such as cyclins, cyclin-dependent kinase inhibitors, I˚B-å and ∫-catenin, for degradation by the proteasome after ubiquitination. Via its F-box domain, FBXO45 can directly interact with Skp1 p19 and CUL-1. FBXO45 has been shown to be an estrogen-induced gene.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FBXO45
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat