Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ATP6V0D2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-ATP6V0D2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-12548R-A555
Supplier:  Bioss
Anti-ATP6V0D2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-12548R-A555
Supplier:  Bioss

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d2
  • Antigen symbol:
    ATP6V0D2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    245972
  • Antigen synonyms:
    VMA6|ATP6D2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    315-350/350
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Subunit of the integral membrane V0 complex of vacuolar ATPase. Vacuolar ATPase is responsible for acidifying a variety of intracellular compartments in eukaryotic cells, thus providing most of the energy required for transport processes in the vacuolar system. May play a role in coupling of proton transport and ATP hydrolysis (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATP6V0D2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat