Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Aspartate Beta-Hydroxylase
- Antigen symbol:ASPH
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CASQ2BP1|Peptide aspartate beta dioxygenase|Aspartyl/asparaginyl beta hydroxylase|BAH|HAAH|ASP beta hydroxylase|JCTN|ASPH_HUMAN.|junctin
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Aspartyl/asparaginyl beta-hydroxylase (ASPH) is a widely-expressed type II membrane protein involved in calcium homeostasis. Located in the endoplasmic reticulum, ASPH specifically hydroxylates an Asp or Asn residue in the epidermal growth factor-like (EGF) domains of several proteins, using iron as a cofactor. The ASPH gene encodes 3 proteins, ASPH, Junctin, and Junctate (or Humbug), that differ significantly in their C-terminal domains. These ASPH gene products are expressed as five transcript variants that differ by their roles in calcium storage and release, hydroxylation capabilities, and tissue specificity. While all ASPH variants are expressed in skeletal muscle, only some are detected in heart, brain, pancreas, placenta, lung, liver, and kidney tissues. In the lumen of the endoplasmic reticulum, ASPH can be processed into two different forms.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ASPH
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat