Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Cyclin-dependent Kinase 17
- Antigen symbol:CDK17
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:PCTAIRE-2|PCTK2|PCTAIRE-mot protein kinase 2|Protein kinase cdc2-related PCTAIRE-2|6430598J10Rik|CDK17_HUMAN.|MGC25109|Serine/threonine-protein kinase PCTAIRE-2|EC 2.7.1.37
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Cell cycle progression is controlled in part by a family of cyclin proteins and cyclin dependent kinases (Cdks). Cdk proteins work in concert with cyclins to phosphorylate key substrates involved in cell cycle progression. Another family of proteins, Cdk inhibitors, also play a role in regulating the cell cycle by binding to cyclin-Cdk complexes and modulating their activity. Members of the Cdk family include Cdk2–Cdk8, PCTAIRE-1–3, PITALRE and PITSLRE. PCTAIRE-1, PCTAIRE-2 and PCTAIRE-3 comprise a subfamily of cdc2-related serine/threonine kinases. PCTAIRE-1, which is expressed primarily in mammalian brain, interacts with a variety of proteins and is thought to be part of a multiple signal transduction cascade. PCTAIRE-2, also expressed in brain, may be important in terminally differentiated neurons.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CDK17
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: