Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Calcium Homeostasis Modulator 1
- Antigen symbol:CALHM1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:FAM26C|Protein FAM26C|CALHM1|Calcium homeostasis modulator protein 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
CALHM1 is a 346 amino acid multi-pass endoplasmic reticulum membrane protein that belongs to the FAM26 family. CALHM1 co-localizes with GRP 78 to the endoplasmic reticulum. Predominantly expressed in adult brain, CALHM1 may be a pore-forming ion channel that controls cytosolic Ca2+ permeability and cytosolic Ca2+ concentration in the cell. It is suggested that CALHM1 regulates amyloid precursor protein proteolysis and aggregated amyloid-beta peptides levels in a Ca2+ dependent manner. CALHM1 homomultimerizes and shares strong sequence similarities with the selectivity filter of the NMDA receptor, which generates a large Ca2+ conductance across the plasma membrane. CALHM1 may be a potential factor involved in the pathogenesis of Alzheimer's disease.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CALHM1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat