Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CDNF Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-CDNF Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-11499R-A555
Supplier:  Bioss
Anti-CDNF Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-11499R-A555
Supplier:  Bioss

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cerebral dopamine neurotrophic factor
  • Antigen symbol:
    CDNF
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    441549
  • Antigen synonyms:
    pOV-2|ARMETL1|PPAT|UKR1|CDNF|NBIA6|DPCK|NBP|Cerebral dopamine neurotrophic factor
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    100-150/187
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Trophic factor for dopamine neurons. Prevents the 6-hydroxydopamine (6-OHDA)-induced degeneration of dopaminergic neurons. When administered after 6-OHDA-lesioning, restores the dopaminergic function and prevents the degeneration of dopaminergic neurons in substantia nigra (By similarity).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDNF
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: