Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Hydroxysteroid (17-beta) Dehydrogenase 8
- Antigen symbol:HSD17B8
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:7923
- Antigen synonyms:FABGL|KE6|D6S2245E|FABG|RING2|HKE6|SDR30C1|dJ1033B10.9|H2-KE6
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
17beta-HSD8 belongs to the 17beta-HSD family of proteins that regulate the availability of steroids within a tissue. 17beta-HSD8 converts active steroids to their inactive form through its oxidative activity. It is a key player in the inactivation of Estradiol and Testosterone. 17beta-HSD8 is predominantly expressed in placenta, endometrium and prostate but can also be found in liver, and pancreas, with lowest levels found in testis, ovary and kidney. It has been proposed that a reduction in the levels of 17beta-HSD8 may lead to abnormal elevations in the local level of sex steroids, which can lead to recessive renal cystic disease. It has also been suggested that low levels of 17beta-HSD proteins may result in an underdeveloped urogenital system.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HSD17B8
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat