Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen name:Ras-related protein Rab-17
- Antigen symbol:RAB17
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:BOFS|AP2TF|AP-2|AP-2alpha|TFAP2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Rab17 belongs to the Rab family of small Ras-like GTPases. It is specifically expressed in epithelial cells and is upregulated during cell polarization. Immunofluorescence staining studies indicate that Rab17 is associated with the perinuclear recycling endosome in nonpolarized epithelial cells and with the apical recycling endosome in polarized epithelial cells. The function of Rab17 remains unclear. Reports of Rab17 colocalization with internalized IgA in the apical endosome suggest that it may regulate receptor-mediated transcytosis. Rab17 has also been shown to regulate melanocytic filopodia formation and melanosome trafficking. siRNA knockdown of Rab17 in melanoma cells induces melanosome accumulation in the cell periphery.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RAB17
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat