ContactUsLinkComponent
Soldering tweezers
Soldering tweezers
Catalog # 232-1919
Supplier:  HAMMACHER KARL
undefined
Soldering tweezers
Catalog # 232-1919
Supplier:  HAMMACHER KARL
Specifications
  • Type:
    Fine, curved tip
  • Length:
    160 mm
  • Pk:
    1
Specifications
About this item
Wironit steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).
  • Resistant to corrosion
  • Anti-magnetic