ContactUsLinkComponent
Earmuffs, helmet-mounted, Endurance, PW47
Earmuffs, helmet-mounted, Endurance, PW47
Catalog # PRTWPW47RER
Supplier:  Portwest
undefined
Earmuffs, helmet-mounted, Endurance, PW47
Catalog # PRTWPW47RER
Supplier:  Portwest
Specifications
  • SNR:
    29 dB
  • Colour:
    Red
  • Description:
    Earmuffs, Endurance, helmet-mounted, for helmets, PS53, PW54, PW55, 29 dB, red
  • Pk:
    1
Specifications
About this item
These helmet-mounted ear protectors made from ABS feature an excellent attenuation level.
  • Can be adjusted into stand-off position when not in use
  • Easy to clip on
Compatible with Portwest hemlets PS53, PW54 and PW55.
SNR: 29 dB
Certifications: EN 352-3