Order Entry
Northern Ireland
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Ethylene oxide solution
Catalog # CRM44609
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Ethylene oxide solution
Catalog # CRM44609
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    500 µg/ml in DMSO
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    75-21-8
  • Description:
    Ethylene oxide solution
  • Environmentally Preferable:
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00014482
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    44,05
  • Flash point:
    87 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

Certifications: Certified in accordance with ISO 17034 and ISO/IEC 17025:2005.