Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-KVbeta.3 Rabbit Polyclonal Antibody [clone: Q327]
Anti-KVbeta.3 Rabbit Polyclonal Antibody [clone: Q327]
Catalog # BWRLBS9168
Anti-KVbeta.3 Rabbit Polyclonal Antibody [clone: Q327]
Catalog # BWRLBS9168

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    KV beta.3
  • Clonality:
    Polyclonal
  • Clone:
    Q327
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 301-350 of Human KVbeta.3.

KV?.3 (Q327) pAb detects endogenous levels of KV?.3 protein.

Type: Primary
Antigen: KVbeta.3
Clonality: Polyclonal
Clone: Q327
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat