Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen symbol:p-PLC gamma1
- Clonality:Polyclonal
- Clone:Y1253
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human PLC gamma1 around the phosphorylation site of Tyrosine 1253.
p-PLC ?1 (Y1253) pAb detects endogenous levels of PLC ?1 protein when phosphorylated at Tyr1253.
Type: Primary
Antigen: p-PLC gamma1
Clonality: Polyclonal
Clone: Y1253
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat