Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen symbol:p-IFN-gammaR alpha
- Clonality:Polyclonal
- Clone:Y457
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human IFN-gammaRalpha around the phosphorylation site of Tyrosine 457.
p-IFN-?R? (Y457) pAb detects endogenous levels of IFN-?R? protein only when phosphorylated at Tyr457.
Type: Primary
Antigen: p-IFN-gammaRalpha
Clonality: Polyclonal
Clone: Y457
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat