Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Desmin Rabbit Polyclonal Antibody [clone: V56]
Anti-Desmin Rabbit Polyclonal Antibody [clone: V56]
Catalog # BWRLBS1860
Anti-Desmin Rabbit Polyclonal Antibody [clone: V56]
Catalog # BWRLBS1860

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Desmin
  • Clonality:
    Polyclonal
  • Clone:
    V56
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 20-70 of Human Desmin.

Desmin (V56) pAb detects endogenous levels of Desmin protein.

Type: Primary
Antigen: Desmin
Clonality: Polyclonal
Clone: V56
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat