Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Chemokine Receptor D6 Rabbit Polyclonal Antibody [clone: S383]
Anti-Chemokine Receptor D6 Rabbit Polyclonal Antibody [clone: S383]
Catalog # BWRLBS2588
Anti-Chemokine Receptor D6 Rabbit Polyclonal Antibody [clone: S383]
Catalog # BWRLBS2588

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Chemokine Receptor D6
  • Clonality:
    Polyclonal
  • Clone:
    S383
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 361-510 of Human Chemokine Receptor D6.

Chemokine Receptor D6 (S383) pAb detects endogenous levels of Chemokine Receptor D6 protein.

Type: Primary
Antigen: Chemokine Receptor D6
Clonality: Polyclonal
Clone: S383
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat