Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-COL3A1 Rabbit Polyclonal Antibody [clone: P104]
Anti-COL3A1 Rabbit Polyclonal Antibody [clone: P104]
Catalog # BWRLBS1531
Anti-COL3A1 Rabbit Polyclonal Antibody [clone: P104]
Catalog # BWRLBS1531

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    COL3A1
  • Clonality:
    Polyclonal
  • Clone:
    P104
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 71-120 of Human COL3A1.

COL3A1 pAb detects endogenous levels of Collagen III protein.

Type: Primary
Antigen: COL3A1
Clonality: Polyclonal
Clone: P104
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat