To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen symbol:Myf-5
- Clonality:Polyclonal
- Clone:A88
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 60-100 of Human Myf-5.
Myf-5 (A88) pAb detects endogenous levels of Myf-5 protein.
Type: Primary
Antigen: Myf-5
Clonality: Polyclonal
Clone: A88
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse