D-(+)-Trehalose, anhydrous, powder
:99-20-7
:
:C₁₂H₂₂O₁₁
:MFCD00006628
:342.3 g/mol
:Ambient
:202-739-6
- Form:Powder
- Solubility:Soluble in water
- Sensitivity:Hygroscopic
- Storage:Store in cool, dry conditions in well sealed containers. Keep container tightly closed
Frequently Bought Together