Poly-D-lysine hydrobromide, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:27964-99-4
:
Formula:C₆H₁₅BrN₂O₂
:227.102 g/mol
Storage Temperature:Freezer
Specification Test Results
- Identity Test:Passes
- Avg. Molecular Weight:70,000-150,000
Specification Test Results
Frequently Bought Together