Order Entry
Northern Ireland
ContactUsLinkComponent
β-(N-Morpholino)ethanesulphonic acid (MES) hydrate ≥99%
Irritant
β-(N-Morpholino)ethanesulphonic acid (MES) hydrate ≥99%
Catalog # H56472.36
CAS Number:  
Irritant
β-(N-Morpholino)ethanesulphonic acid (MES) hydrate ≥99%
Catalog # H56472.36
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Appearance (colour):
    White
  • Assay:
    99.0%
  • Water Content (Karl Fischer Titration)::
    8.0% to 9.0%

Specification Test Results

About this item

 :
 :195.24 g/mol (anhy)
Merck Index:14,5902
Melting Pt:∼ 308 °C (dec.)
EINECS:224-632-3
 :GHS07
Storage Temperature:Ambient
CAS Number:1266615-59-1
Formula:C₆H₁₃NO₄S·xH₂O
MDL Number:MFCD00149409