Order Entry
Northern Ireland
ContactUsLinkComponent
Ethyl-(S)-(-)-lactate, EMPLURA®
Flammable
Corrosive
Irritant
Ethyl-(S)-(-)-lactate, EMPLURA®
Catalog # 1.09639.2500
Supplier:  Merck
CAS Number:  
Flammable
Corrosive
Irritant
Ethyl-(S)-(-)-lactate, EMPLURA®
Catalog # 1.09639.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC, area%):
    ≥ 99.0 % (a/a)
  • Density (20/4):
    1.032 - 1.034
  • Refreactive index (n25/d):
    1.410 - 1.420
  • Water (KF):
    ≤ 0.20 %
  • Identity (IR):
    passes test

Specification Test Results

About this item

 :
 :118.13 g/mol
Melting Pt:–25 °C
EINECS:211-694-1
ADR:3,
 :M-Clarity™ quality level = MQ200
Flash Pt:53 °C
UN:1192
 :GHS02
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:687-47-8
 :MFCD00065359
Formula:C₅H₁₀O₃
MDL Number:MFCD00004518