Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Keratin 7
- Antigen symbol:KRT7
- Clonality:Monoclonal
- Clone:rOV-TL12/30
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:3855
- Storage buffer:In 10 mM PBS (0.05% BSA and 0.05% azide).
- Storage temperature:Store at 4° C.
- Immunogen:OTN 11 ovarian carcinoma cell line.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal antibody raised against human KRT7.
Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: rOV-TL12/30
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human