Ground protection systems, steel
Supplier: CEMO
Certificates
About this item
These ground protection systems ensures safe handling of water-polluting as well as flammable substances - previously classed as AI*, AII, AIII and B.
- Storage of water-polluting and flammable substances
- Painted steel RAL 5002, ultramarine blue
- Hot-dip galvanised gratings
Modular matched elements enable the lining of any large surfaces as a storage or transfilling station.
Frequently Bought Together