Order Entry
Northern Ireland
ContactUsLinkComponent
Human ICAM-2 ELISA kit
Human ICAM-2 ELISA kit
  BIRBORB138100-96
 :  Biorbyt
 :  
undefined
Human ICAM-2 ELISA kit
  BIRBORB138100-96
 :  Biorbyt
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Target protein:
    ICAM-2
  • Description:
    Human ICAM-2 ELISA Kit
  • Detection range:
    312 - 20000 pg/ml
  • Sensitivity:
    <10 pg/ml

 

 

Human ICAM-2 ELISA kit for the quantitative detection of human Intercellular adhesion molecule 2 (ICAM-2) in cell culture supernates, tissue homogenates, serum and plasma (heparin, EDTA).

ICAM2 also known as CD102 is an integral membrane protein that has 2 immunoglobulin-like domains. This gene is mapped to 17q23.3. The protein encoded by this gene is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. This protein may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance.