Order Entry
Northern Ireland
ContactUsLinkComponent
Mouse recombinant mMIP 1a (from E. coli)
Mouse recombinant mMIP 1a (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse recombinant mMIP 1a (from E. coli)
Supplier:  Biorbyt

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Lyophilized MIP-1a although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution CCL3 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Biological activity:
    The Activity is calculated by the ability to chemoattract of Balb3/C splenocytes using 1-10 ng/ml.
  • Protein synonyms:
    Small inducible cytokine A3
  • Protein/peptide name:
    mMIP 1a
  • Purity:
    Greater than 99.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Macrophage Inflammatory Protein-1a in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    The sequence of the first five N-terminal amino acids was determined and was found to be, Ala-Pro-Tyr-Gly-Ala.
  • Formulation:
    The protein was lyophilized from 1mg/ml solution containing no additives.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together