IFN alpha 2B
Catalog # BIRBORB97779-100
Supplier: Biorbyt
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:100 µG
- Storage conditions:Store at−20 °C.
- Biological activity:EC50: 2.6x108 IU/mg, determined in a viral resistanceassay using bovine kidney MDBK cells.
- UniProtKB:Q86UP4
- Protein/peptide name:IFN alpha 2B
- Purity:95% pure by SDS-PAGE and RP-HPLC
- Solubility:It is recommended to reconstitute the lyophilized IFN- insterile bidest water not less than 100 g/ml, which canthen be further diluted to other aqueous solutions. Forlong term storage it is recommended to add a carrierprotein (0.1% HSA or BSA).
- Molecular weight:19.4 kDa
- Formulation:Lyophilized.Lyophilized from a (1 mg/ml) solution containing 2.3 mg Sodium phosphate dibasic and 0.55 mgsodium phosphate monobasic buffer.