Mouse FGF basic (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Source:E. coli
- Species:Mouse
- Storage conditions:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Biological activity:The ED50 range= 0.1ng/ml, determined by the dose-dependant proliferation of BALB/3T3 cells.
- Protein synonyms:HBGH-2
- Protein/peptide name:FGF basic
- Purity:> 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:It is recommended to reconstitute the lyophilized Fibroblast Growth Factor in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Sequence:The sequence of the first six N-terminal amino acids was determined and was found to be Met-Pro-Ala-Leu-Pro-Glu.
- Formulation:FGF-b was lyophilized from 5mM NaP, pH7.5 and 50mM NaCl.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together