Bottles, narrow neck, with screw cap
Supplier: witeg Labortechnik
Certificates
About this item
Narrow neck bottles, made of soda glass, amber, with black screw cap.
- Good chemical and physical properties
- For applications that do not involve high thermal stresses
- Round thread in accordance with DIN 168
Excellent for sampling, storing and transporting liquids. Amber coloured to protect light-sensitive materials.
Frequently Bought Together