Specifications
- Pk:20 Tests
- Description:Universal methylated DNA standard (human)
Specifications
About this item
Universal methylated DNA standard (human) with the included primers can be used in conjunction with Enzo blood and tissue methylation kit or the express methylation kits.
- Convenient for use with range of Methylation Kits
- Enzymatically modified standard includes short, easy to follow protocol
- Primers included
The DNA included in this set is enzymatically methylated at all cytosine positions to help assess the efficiency of the bisulfite-mediated conversion of DNA. DNA is isolated from a male human source. The primer set is designed to amplify a fragment of the human mismatch MLH1 mismatch repair gene following bisulfite treatment. The methylated cytosines comprising CG dinucleotides remain unconverted following bisulfite treatment, whereas non-methylated cytosines are converted into uracil and detected as thymine after PCR.