Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-UGT1A9 Mouse Monoclonal Antibody (CF568) [clone: UGT1A9/1229]
Anti-UGT1A9 Mouse Monoclonal Antibody (CF568) [clone: UGT1A9/1229]
Supplier:  Biotium
Certificates
Anti-UGT1A9 Mouse Monoclonal Antibody (CF568) [clone: UGT1A9/1229]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC681229-500
  • BNC681229-100
  • Antigen symbol:
  • UGT1A9
  • UGT1A9
  • Antigen name:
  • UDP glucuronosyltransferase 1 family, polypeptide A9
  • UDP glucuronosyltransferase 1 family, polypeptide A9
  • Antigen synonyms:
  • UDPGT 1-9|UGT1-09|Ugt1a9|UDP-glucuronosyltransferase 1A9 (UGT1A9)|UGT-1I|GNT1|UDP-glucuronosyltransferase 1-I|lugP4|UGT1.9|HLUGP4|UGT1*9|UGT1I|UDPGT
  • UDPGT 1-9|UGT1-09|Ugt1a9|UDP-glucuronosyltransferase 1A9 (UGT1A9)|UGT-1I|GNT1|UDP-glucuronosyltransferase 1-I|lugP4|UGT1.9|HLUGP4|UGT1*9|UGT1I|UDPGT
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone UGT1A9/1229
  • Clone UGT1A9/1229
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 554822
  • 554822
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together