Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-ITGB5 Rabbit Polyclonal Antibody
Anti-ITGB5 Rabbit Polyclonal Antibody
Catalog # ABGEAP14000A
Supplier:  Abgent
Anti-ITGB5 Rabbit Polyclonal Antibody
Catalog # ABGEAP14000A
Supplier:  Abgent

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Integrin Beta 5
  • Antigen symbol:
    ITGB5
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    RGD1563276
  • Size:
    100 µg
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ITGB5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human