Anti-HLA-A; HLA-B; HLA-C Mouse Monoclonal Antibody (CF405S) [clone: 246-B8.E7]
Supplier: Biotium
Certificates
About this item
Reacts with a monomorphic determinant of human major histocompatibility (MHC) class I antigens (HLA-A, B and C). Human MHC class I antigens are expressed constitutively on all nucleated cells lymphocytes such as lymphocytes, thymocytes, granulocytes, and bone marrow cells and are absent on erythrocytes. MHC class I antigens play a role in class I MHC-associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation.
Type: Primary
Antigen: HLA-A; HLA-B; HLA-C
Clonality: Monoclonal
Clone: 246-B8.E7
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Mouse, Non-human primates
Specifications
- Catalog No:
- BNC041054-500
- BNC041054-100
- Antigen symbol:
- MHC I
- MHC I
- Antigen name:
- MHC Class 1
- MHC Class 1
- Antigen synonyms:
- HLA-B|HLA-A|HLA-C
- HLA-B|HLA-A|HLA-C
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 405S
- CF® 405S
- Clone:
- Clone 246-B8.E7
- Clone 246-B8.E7
- Reactivity:
- Primates, Human, Mouse
- Primates, Human, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 181244; 654404; 77961
- 181244; 654404; 77961
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together