Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CD74 Mouse Monoclonal Antibody
Anti-CD74 Mouse Monoclonal Antibody
  NOVUNB100-1985
Anti-CD74 Mouse Monoclonal Antibody
  NOVUNB100-1985

 

  • Antibody type:
    Primary
  • Antigen symbol:
    CD74
  • Clonality:
    Monoclonal
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 μg
  • Pk:
    0,1 mL

 

 

Mouse Monoclonal CD74 Antibody (PIN.1). Tested Applications: Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation. Tested Reactivity: Human.

Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: