About this item
Glypican-3 (GPC3) is an integral membrane protein that is mutated in the Simpson-Golabi-Behmel syndrome (SGBS). SGBS is characterized by pre- and post-natal overgrowth and is a recessive X-linked condition.GPC3 may also be found in a secreted form. Anti-GPC3 has been identified as a useful tumor marker for the diagnosis of hepatocellular carcinoma (HCC), hepatoblastoma, melanoma, testicular germ cell tumors, and Wilm s tumor. In patients with HCC, GPC3 is overexpressed in neoplastic liver tissue and elevated in serum, but is undetectable in normal liver, benign liver, and the serum of healthy donors. GPC3 expression is also found to be higher in HCC liver tissue than in cirrhotic liver or liver with focal lesions such as dysplastic nodules and areas of hepatic adenoma (HA) with malignant transformation. In the context of testicular germ cell tumors, GPC3 expression is up regulated in certain histologic subtypes, specifically yolk sac tumors and choriocarcinoma. A high level of GPC3 expression has also been found in some types of embryonal tumors, such as Wilm s tumor and hepatoblastoma, with a low or undetectable expression in normal adjacent tissue. In patients with thyroid cancer, expression of GPC3 is dramatically enhanced in certain types of cancers: 100% in follicular carcinoma and 70% in papillary carcinoma. Expression of GPC3 in follicular carcinoma was significantly higher than that of follicular adenoma. In contrast, GPC 3 is not expressed in anaplastic carcinoma.
Type: Primary
Antigen: GPC3
Clonality: Monoclonal
Clone: GPC3/863 1G12
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Human
Specifications
- Catalog No:
- BNC470864-100
- BNC470864-500
- Antigen symbol:
- GPC3
- GPC3
- Antigen name:
- Glypican 3
- Glypican 3
- Antigen synonyms:
- GPC3|Heparan sulphate proteoglycan|SDYS|MXR7|SGBS1|OCI-5|Secreted glypican-3|GTR2-2|Glypican proteoglycan 3|DGSX|Intestinal protein OCI-5
- GPC3|Heparan sulphate proteoglycan|SDYS|MXR7|SGBS1|OCI-5|Secreted glypican-3|GTR2-2|Glypican proteoglycan 3|DGSX|Intestinal protein OCI-5
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- Clone GPC3/863 1G12
- Clone GPC3/863 1G12
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 644108
- 644108
- Isotype:
- IgGs
- IgGs
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide