Anti-KRT7 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: OV-TL12/30]
Catalog # BNUM0683-50
Supplier: Biotium
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 7
- Antigen symbol:KRT7
- Clonality:Monoclonal
- Clone:Clone OV-TL12/30
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Gene ID:411501
- Antigen synonyms:KRT7|Krt2-7|Sarcolectin|SCL|K2C7|Keratin 55K Type II Cytoskeletal|CK-7|Type-II Keratin Kb7|Type II Mesothelial Keratin K7|Keratin Simple Epithelial Type 1 K7|Keratin Type II Cytoskeletal 7
- Storage buffer:PBS/0,05% azide
- Concentration:1 mg/mL
- Purification:BSA-free
- Size:50 μL
- Pk:50 µl
Specifications
About this item
This antibody recognizes an intermediate filament protein (IFP) of 55 kDa, which is identified as cytokeratin 7. This MAb is highly specific to cytokeratin 7 and shows no cross-reaction with other IFPs. Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. This MAb is highly useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).
Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: OV-TL12/30
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human