Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CD79A Mouse Monoclonal Antibody (Biotin) [clone: HM47/A9]
Anti-CD79A Mouse Monoclonal Antibody (Biotin) [clone: HM47/A9]
Supplier:  Biotium
Certificates
Anti-CD79A Mouse Monoclonal Antibody (Biotin) [clone: HM47/A9]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB0477-100
  • BNCB0477-500
  • Antigen symbol:
  • CD79A
  • CD79A
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone HM47/A9
  • Clone HM47/A9
  • Reactivity:
  • Pig, Monkey, Human, Bovine, Rat, Mouse
  • Pig, Monkey, Human, Bovine, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 631567
  • 631567
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together