Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-NEFH Mouse Monoclonal Antibody (CF488A) [clone: RT-97]
Anti-NEFH Mouse Monoclonal Antibody (CF488A) [clone: RT-97]
Supplier:  Biotium
Certificates
Anti-NEFH Mouse Monoclonal Antibody (CF488A) [clone: RT-97]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC880454-100
  • BNC880454-500
  • Antigen symbol:
  • NEFH
  • NEFH
  • Antigen name:
  • Neurofilament, Heavy Polypeptide
  • Neurofilament, Heavy Polypeptide
  • Antigen synonyms:
  • Neurofilament H|Neurofilament Heavy Polypeptide 200kDa|NEFH|NF-H|Neurofilament Triplet H Protein|NF200
  • Neurofilament H|Neurofilament Heavy Polypeptide 200kDa|NEFH|NF-H|Neurofilament Triplet H Protein|NF200
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone RT-97
  • Clone RT-97
  • Reactivity:
  • Pig, Chicken, Human, Rat, Mouse
  • Pig, Chicken, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 198760
  • 198760
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together