Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-HSPG2 Rat Monoclonal Antibody (CF647) [clone: A7L6]
 :  Biotium
 
Anti-HSPG2 Rat Monoclonal Antibody (CF647) [clone: A7L6]
 :  Biotium

 

  • Catalog No:
  • BNC470315-500
  • BNC470315-100
  • Antigen symbol:
  • HSPG2
  • HSPG2
  • Antigen name:
  • Heparan Sulphate Proteoglycan
  • Heparan Sulphate Proteoglycan
  • Antigen synonyms:
  • LG3 peptide|HSPG|PLC|Perlecan|Heparan sulfate proteoglycan of basement membrane|Hspg2|SJS1
  • LG3 peptide|HSPG|PLC|Perlecan|Heparan sulfate proteoglycan of basement membrane|Hspg2|SJS1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone A7L6
  • Clone A7L6
  • Reactivity:
  • Fish, Pig, Monkey, Human, Bovine, Mouse
  • Fish, Pig, Monkey, Human, Bovine, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 562227
  • 562227
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together