About this item
This MAb specifically precipitates heterogeneous material of high MW, identified as perlecan, a major heparan-sulfate proteoglycan (HSPG) within all basement membranes and cell surfaces. It does not cross-react with laminin, fibronectin, or dermatran sulfate proteoglycan. Because of perlecan s strategic location and ability to store and protect growth factors, it has been strongly implicated in the control of tumor cell growth and metastatic behavior. Perlecan possesses angiogenic and growth-promoting attributes primarily by acting as a co-receptor for basic fibroblast growth factor (FGF-2). Suppression of perlecan causes substantial inhibition of neoplastic growth and neovascularization. Thus, perlecan is a potent inducer of neoplasm growth and angiogenesis in vivo and therapeutic interventions targeting this key modulator of tumor progression may improve neoplastic treatment.
Type: Primary
Antigen: HSPG2
Clonality: Monoclonal
Clone: A7L6
Conjugation: CF488A
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Cow, Fish, Human, Monkey, Mouse, Pig
Specifications
- Catalog No:
- BNC880315-100
- BNC880315-500
- Antigen symbol:
- HSPG2
- HSPG2
- Antigen name:
- Heparan Sulphate Proteoglycan
- Heparan Sulphate Proteoglycan
- Antigen synonyms:
- LG3 peptide|HSPG|PLC|Perlecan|Heparan sulfate proteoglycan of basement membrane|Hspg2|SJS1
- LG3 peptide|HSPG|PLC|Perlecan|Heparan sulfate proteoglycan of basement membrane|Hspg2|SJS1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone A7L6
- Clone A7L6
- Reactivity:
- Fish, Pig, Monkey, Human, Bovine, Mouse
- Fish, Pig, Monkey, Human, Bovine, Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 562227
- 562227
- Isotype:
- IgG2a kappa
- IgG2a kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide