About this item
Recognizes a protein of 110 kDa, identified as CD106 (also known as vascular cell adhesion molecule-1 (VCAM-1) and INCAM-100). CD106 is a member of the Ig superfamily of adhesion molecules and is expressed at high levels on cytokine stimulated vascular endothelial cells, and at minimal levels on un-stimulated endothelial cells. It is also present on follicular and inter-follicular dendritic cells of lymph nodes, myoblasts, and some macrophages. CD106 serves as a ligand for leukocyte integrin (VLA-4 or CD49d/CD29) and mediates cell adhesion of leukocytes to activated endothelium. It plays a role in various immunological and inflammatory responses. This MAb inhibits the binding of leukocytes to VCAM-1 on stimulated endothelial cells.
Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: B-K9
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC470304-500
- BNC470304-100
- Antigen symbol:
- VCAM1
- VCAM1
- Antigen name:
- Vascular cell adhesion molecule 1
- Vascular cell adhesion molecule 1
- Antigen synonyms:
- CD106|Vascular cell adhesion protein 1|INCAM-100|Vascular Cell Adhesion Molecule 1 (VCAM1)|L1CAM
- CD106|Vascular cell adhesion protein 1|INCAM-100|Vascular Cell Adhesion Molecule 1 (VCAM1)|L1CAM
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- Clone B-K9
- Clone B-K9
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 109225
- 109225
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide