Anti-HLA-DPB1 Mouse Monoclonal Antibody (CF488A) [clone: BRA-FB6]
Supplier: Biotium
Certificates
About this item
Recognizes a non-polymorphic determinant of DP-MHC class II. MHC class II antigens are transmembrane glycoproteins of non-covalently linked α (33-35 kDa) and β (27-30 kDa) chains. It reportedly reacts with B- & non-T, non-B cell lines but not with T- and myeloid cell lines and leukemias. Differential expression of MHC class II antigens on fetal and adult lymphocytes, malignant B cells appears to reflect the stage of cell differentiation which may be useful in the study of lymphoproliferative disorders.
Type: Primary
Antigen: HLA-DPB1
Clonality: Monoclonal
Clone: BRA-FB6
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC880199-100
- BNC880199-500
- Antigen symbol:
- MHC2
- MHC2
- Antigen name:
- MHC II
- MHC II
- Antigen synonyms:
- HLA-DPB1
- HLA-DPB1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 488A
- CF® 488A
- Clone:
- Clone BRA-FB6
- Clone BRA-FB6
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 347270
- 347270
- Isotype:
- IgG2b kappa
- IgG2b kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together