About this item
Cytokeratin 17 (CK17) is normally expressed in the basal cells of complex epithelia but not in stratified or simple epithelia. Antibody to CK17 is an excellent tool to distinguish myoepithelial cells from luminal epithelium of various glands such as mammary, sweat and salivary. CK17 is expressed in epithelial cells of various origins, such as bronchial epithelial cells and skin appendages. It may be considered as anepithelial stem cellmarker because CK17 Ab marks basal cell differentiation. CK17 is expressed in SCLC much higher than in LADC. Eighty-five percent of the triple negative breast carcinomas immunoreact with basal cytokeratins including anti-CK17. Also important is that cases of triple negative breast carcinoma with expression of CK17 show an aggressive clinical course. The histologic differentiation of ampullary cancer, intestinal vs. pancreatobiliary, is very important for treatment. Usually anti-CK17 and anti-MUC1 immunoreactivity represents pancreatobiliary subtype whereas anti-MUC2 and anti-CDX-2 positivity defines intestinal subtype.
Type: Primary
Antigen: KRT17
Clonality: Monoclonal
Clone: E3
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Cow, Goat, Human, Pig, Rat
Specifications
- Catalog No:
- BNC470158-500
- BNC470158-100
- Antigen symbol:
- KRT17
- KRT17
- Antigen name:
- Cytokeratin 17
- Cytokeratin 17
- Antigen synonyms:
- Keratin-17|KRT17|Keratin Type I Cytoskeletal 17|PCHC1|K17
- Keratin-17|KRT17|Keratin Type I Cytoskeletal 17|PCHC1|K17
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 647
- CF® 647
- Clone:
- Clone E3
- Clone E3
- Reactivity:
- Pig, Goat, Human, Bovine, Rat
- Pig, Goat, Human, Bovine, Rat
- Host:
- Mouse
- Mouse
- Gene ID:
- 2785
- 2785
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide