Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-IGHA1 & IGHA2 Mouse Monoclonal Antibody (CF647) [clone: IA-HISA43]
Anti-IGHA1 & IGHA2 Mouse Monoclonal Antibody (CF647) [clone: IA-HISA43]
Supplier:  Biotium
Certificates
Anti-IGHA1 & IGHA2 Mouse Monoclonal Antibody (CF647) [clone: IA-HISA43]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470148-100
  • BNC470148-500
  • Antigen symbol:
  • IGHA1 & IGHA2
  • IGHA1 & IGHA2
  • Antigen name:
  • IgA Immunoglobulin
  • IgA Immunoglobulin
  • Antigen synonyms:
  • Ig alpha 1 Chain C Region|Immunoglobulin Heavy Constant Alpha 2|IGHA2|Ig alpha 2 Chain C Region|Immunoglobulin Heavy Constant Alpha 1|Immunoglobulin Am1|IGHA1|Immunoglobulin Am2|A2m Marker
  • Ig alpha 1 Chain C Region|Immunoglobulin Heavy Constant Alpha 2|IGHA2|Ig alpha 2 Chain C Region|Immunoglobulin Heavy Constant Alpha 1|Immunoglobulin Am1|IGHA1|Immunoglobulin Am2|A2m Marker
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone IA-HISA43
  • Clone IA-HISA43
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 699841
  • 699841
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together