Anti-TAG-72 / CA72.4 Mouse Monoclonal Antibody (CF568) [clone: CA72/145]
Supplier: Biotium
Certificates
About this item
Recognizes an oncofetal antigen of 220 kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. The majority of human adenocarcinomas including colorectal, pancreatic, gastric, ovarian, endometrial, mammary, and non-small cell lung cancer display some cell populations that are positive for TAG72. About 60% of carcinoma patients express TAG72 in their sera. TAG72 is reportedly useful in distinguishing pulmonary adenocarcinomas from pleural mesotheliomas.
Type: Primary
Antigen: TAG-72 / CA72.4
Clonality: Monoclonal
Clone: CA72/145
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Specifications
- Catalog No:
- BNC680145-100
- BNC680145-500
- Antigen name:
- TAG-72 / CA72.4
- TAG-72 / CA72.4
- Antigen synonyms:
- CA72.4|Tumor associated glycoprotein 72
- CA72.4|Tumor associated glycoprotein 72
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone CA72/145
- Clone CA72/145
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide
Specifications
Frequently Bought Together