Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CD33 Mouse Monoclonal Antibody (Biotin) [clone: C33/69]
Anti-CD33 Mouse Monoclonal Antibody (Biotin) [clone: C33/69]
Supplier:  Biotium
Certificates
Anti-CD33 Mouse Monoclonal Antibody (Biotin) [clone: C33/69]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNCB0069-100
  • BNCB0069-500
  • Antigen symbol:
  • CD33
  • CD33
  • Antigen synonyms:
  • gp67|Sialic acid-binding Ig-like lectin 3 (SIGLEC3)|My9|Myeloid cell surface antigen CD33|p67
  • gp67|Sialic acid-binding Ig-like lectin 3 (SIGLEC3)|My9|Myeloid cell surface antigen CD33|p67
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Clone:
  • Clone C33/69
  • Clone C33/69
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 83731
  • 83731
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together