Oil, for heat transfer
MDL No.: MFCD00147787
Density: 0,86
Melting point: –6 °C
Flash point: 230 °C
Appearance: Clear light yellow oily liquid
Storage temperature: Ambient
- Melting point:-6°C
- Density:0.86
- Flash point:230°C
Frequently Bought Together