Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-APP Mouse Monoclonal Antibody [clone: 3E9]
Anti-APP Mouse Monoclonal Antibody [clone: 3E9]
Supplier:  ENZO LIFE SCIENCES
Certificates
Anti-APP Mouse Monoclonal Antibody [clone: 3E9]
Supplier:  ENZO LIFE SCIENCES

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Catalog No:
  • ENZOADINBA100E
  • Antigen symbol:
  • APP
  • Antigen name:
  • Amyloid beta (A4) Precursor Protein
  • Antigen synonyms:
  • CTFgamma|CVAP|ABETA|APPI|AD1|PN-II|ABPP|AAA|PN2
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • 3E9
  • Reactivity:
  • Human, Mouse
  • Host:
  • Mouse
  • Isotype:
  • IgG1
  • Western blot:
  • Yes
  • Immunogen:
  • Synthetic peptide corresponding to a portion of human APP695
  • Form:
  • Liquid
  • Purification:
  • Purified by Protein A affinity chromatography
  • Storage buffer:
  • PBS, pH 7,2 containing 50% glycerol
  • Storage temperature:
  • −20 °C
  • Shipping temperature:
  • Blue ice not frozen

Specifications

Frequently Bought Together