Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Antibody type:Primary
- Antigen symbol:HA-Tag
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Form:Liquid
- Storage buffer:PBS, pH 7,2, containing 50% glycerol.
- Sequence:YPYDVPDYA
- Storage temperature:–20 °C
- Shipping temperature:Shipped on Blue Ice
- Immunogen:HA-tag (YPYDVPDYA) conjugated to KLH.
- Purification:purified by Protein A affinity chromatography
- Size:100 μL
- Pk:100 µl
The Ha-Tag (YPYDVPDYA) corresponds to the partial peptide sequence of the influenza hemagglutinin protein. Recognizes the HA-tag of HA-tagged recombinant proteins.
Primary: Tag
Antigen: HA-tag
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: